Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253681.1 | NP_001240610.1 | 15 | 1914 | ||
NM_001253686.1 | NP_001240615.1 | 14 | 1899 | |||
NM_001253687.1 | NP_001240616.1 | 13 | 1803 | |||
NM_001253688.1 | NP_001240617.1 | 13 | 1713 | |||
NM_001253689.1 | NP_001240618.1 | 13 | 1684 | |||
NM_029673.3 | NP_083949.2 | 15 | 1947 | |||
GenBank mRNA | AK006977.1 | - | 3 | 292 | ||
AK033126.1 | - | 13 | 1714 | |||
AK038129.1 | - | 15 | 1937 | |||
AK049189.1 | - | 15 | 1858 | |||
AK146640.1 | - | 13 | 1676 | |||
AK152588.1 | - | 14 | 1845 | |||
AK159909.1 | - | 14 | 1850 | |||
AK163491.1 | - | 15 | 1822 | |||
AK169365.1 | - | 13 | 1616 | |||
BC025034.1 | - | 3 | 692 | |||
BC055840.1 | - | 3 | 376 | |||
BC061010.1 | - | 4 | 418 | |||
BC080790.1 | - | 2 | 264 | |||
BC108356.1 | - | 7 | 845 | |||
BC145704.1 | - | 2 | 227 | |||
BC145706.1 | - | 2 | 227 |