Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165991.1 | NP_001159463.1 | 5, 6 | 553 | ||
NM_001165992.1 | NP_001159464.1 | 5, 6 | 534 | |||
NM_001165995.1 | NP_001159467.1 | 4, 5 | 440 | |||
NM_029702.4 | NP_083978.3 | 5, 6 | 547 | |||
GenBank mRNA | AJ413953.1 | - | 5, 6 | 500 | ||
AK005174.1 | - | 5, 6 | 495 | |||
AK044759.1 | - | 5, 6 | 505 | |||
AK050136.1 | - | 2, 3 | 1405 | |||
AK185500.1 | - | 2, 3 | 153 | |||
BC010713.1 | - | 5, 6 | 494 | 2649462 |