Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029775.2 | NP_084051.1 | 2, 3 | 417 | ||
GenBank mRNA | AK012764.1 | - | 2, 3 | 430 | ||
AK013933.1 | - | 2, 3 | 435 | |||
AK014737.1 | - | 2, 3 | 459 | |||
AK134862.1 | - | 464 | ||||
BC020048.1 | - | 2, 3 | 453 | |||
BC020089.1 | - | 2, 3 | 417 | 4013755 | ||
BC049641.1 | - | 606 |