Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029787.2 | NP_084063.1 | 2, 3 | 237 | ||
GenBank mRNA | AF332059.1 | - | 2, 3 | 161 | ||
AF332060.1 | - | 2, 3 | 161 | |||
AK002640.1 | - | 2, 3 | 209 | |||
AK010858.1 | - | 2, 3 | 214 | |||
AK128917.1 | - | 2, 3 | 231 | |||
AK177067.1 | - | 1, 2 | 67 | |||
AK202011.1 | - | 1, 2 | 67 | |||
AK208840.1 | - | 1, 2 | 67 | |||
BC004760.1 | - | 2, 3 | 244 | 3154531 | ||
BC032013.1 | - | 2, 3 | 219 | 4238275 | ||
BC043074.1 | - | 2, 3 | 237 | 6404746 |