Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029793.2 | NP_084069.1 | 5, 6 | 643 | ||
GenBank mRNA | AK002954.1 | - | 5, 6 | 587 | ||
AK008972.1 | - | 5, 6 | 543 | |||
AK028516.1 | - | 5, 6 | 562 | |||
AK044987.1 | - | 5, 6 | 591 | |||
BC024637.1 | - | 5, 6 | 565 | |||
BC037037.1 | - | 1, 2 | 80 | |||
BC043090.1 | - | 5, 6 | 508 | 6418353 |