Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000351.5 | NP_000342.2 | 4 | 622 | ||
NM_001320750.1 | NP_001307679.1 | 5 | 846 | |||
NM_001320751.1 | NP_001307680.1 | 6 | 840 | |||
NM_001320752.1 | NP_001307681.1 | 5 | 686 | |||
NM_001320753.1 | NP_001307682.1 | 4 | 464 | |||
NM_001320754.1 | NP_001307683.1 | 4 | 612 | |||
GenBank mRNA | AK314034.1 | - | 4 | 562 | ||
AL549736.3 | - | 4 | 622 | |||
BC075030.2 | - | 4 | 398 | 30915417 | ||
J04964.1 | - | 4 | 580 | |||
M16505.1 | - | 4 | 597 |