Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000355.3 | NP_000346.2 | 3 | 665 | ||
GenBank mRNA | AK313711.1 | - | 3 | 555 | ||
BC001176.1 | - | 3 | 543 | 3353271 | ||
CR456591.1 | - | 3 | 455 | |||
L02647.1 | - | 3 | 513 | |||
L02648.1 | - | 3 | 469 | |||
M60396.1 | - | 3 | 453 |