Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285917.1 | NP_001272846.1 | 23 | 3000 | ||
NM_029653.3 | NP_083929.2 | 23 | 3000 | |||
NM_134062.2 | NP_598823.1 | 23 | 2885 | |||
GenBank mRNA | AK145922.1 | - | 23 | 2886 | ||
AY245540.2 | - | 23 | 2993 | |||
AY245541.1 | - | 23 | 2986 | |||
BC021490.1 | - | 21 | 2484 | |||
BC037483.1 | - | 3 | 508 | |||
X97048.1 | - | 22 | 2835 |
Set Membership: |
G-protein coupled receptor |