Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146119.1 | NP_001139591.1 | 11 | 1104 | ||
NM_029758.4 | NP_084034.2 | 12 | 1177 | |||
GenBank mRNA | AK010821.1 | - | 10 | 1040 | ||
AK047142.1 | - | 11 | 1067 | |||
BC049664.1 | - | 12 | 1139 | 6532813 | ||
BC052465.1 | - | 11 | 1085 | 6834768 |