Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029770.2 | NP_084046.2 | 17 | 3586 | ||
GenBank mRNA | AJ487853.1 | - | 17 | 3489 | ||
AK018177.1 | - | 17 | 3586 | |||
AK147603.1 | - | 16 | 3448 | |||
AK154271.1 | - | 17 | 3586 | |||
AK154287.1 | - | 17 | 3591 | |||
AK158284.1 | - | 17 | 3586 | |||
BC048162.1 | - | 16 | 3360 | |||
BC057560.1 | - | 16 | 3360 | 6417563 |