Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029788.5 | NP_084064.1 | 7 | 1126 | ||
GenBank mRNA | AK002624.1 | - | 7 | 1084 | ||
AK036401.1 | - | 7 | 1127 | |||
AK153293.1 | - | 7 | 1095 | |||
AK171077.1 | - | 6 | 2377 | |||
BC037057.1 | - | 6 | 913 | |||
BC050796.1 | - | 7 | 1094 | 6743413 |