Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029802.4 | NP_084078.3 | 5, 6 | 762 | ||
GenBank mRNA | AK009100.1 | - | 5, 6 | 700 | ||
AK154772.1 | - | 5, 6 | 728 | |||
BC013794.1 | - | 5, 6 | 673 | 2648132 | ||
BC022942.1 | - | 5, 6 | 686 | 3583149 | ||
BC028310.1 | - | 5, 6 | 677 | |||
BC034520.1 | - | 5, 6 | 743 | 3964827 | ||
BC079603.1 | - | 2, 3 | 1935 | |||
BU583325.1 | - | 1, 2 | 217 |