Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 4 | 563 | ||
NM_001205392.1 | NP_001192321.1 | 4 | 563 | |||
NM_029846.4 | NP_084122.2 | 4 | 563 | |||
GenBank mRNA | AB087879.1 | - | 4 | 374 | ||
AB087880.1 | - | 4 | 374 | |||
AB087881.1 | - | 4 | 374 | |||
AB476646.1 | - | 4 | 374 | |||
AK005181.1 | - | 4 | 488 | |||
AK030983.1 | - | 4 | 504 | |||
AK048873.1 | - | 4 | 482 | |||
AK131120.1 | - | 4 | 462 | |||
AK134333.1 | - | 4 | 519 | |||
AK145673.1 | - | 4 | 501 | |||
AK155358.1 | - | 4 | 499 | |||
AK170073.1 | - | 4 | 497 | |||
BC049122.1 | - | 4 | 488 | 6813377 |