Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029847.4 | NP_084123.2 | 8 | 2500 | ||
GenBank mRNA | AK013194.2 | - | 8 | 2455 | ||
AK031147.1 | - | 8 | 2488 | |||
AK032812.1 | - | 8 | 2466 | |||
AK035464.1 | - | 8 | 2421 | |||
AK083188.1 | - | 8 | 2454 | |||
AK084090.1 | - | 8 | 2486 | |||
AK086667.1 | - | 8 | 2459 | |||
AK144817.1 | - | 8 | 2468 | |||
AK165970.1 | - | 8 | 2468 |