Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161628.1 | NP_001155100.1 | 3, 4 | 547 | ||
NM_029946.4 | NP_084222.4 | 20, 21 | 2654 | |||
GenBank mRNA | AK019850.1 | - | 3, 4 | 547 | ||
AK220412.1 | - | 3, 4 | 466 | |||
BC056955.1 | - | 8, 9 | 1057 | 6812632 | ||
BC065111.1 | - | 20, 21 | 2654 | 30536704 |