Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030116.2 | NP_084392.1 | 7 | 1186 | ||
GenBank mRNA | AK020217.1 | - | 2 | 529 | ||
AK040055.1 | - | 7 | 1187 | |||
AK046099.1 | - | 7 | 1186 | |||
AK161563.1 | - | 7 | 1190 | |||
AK162023.1 | - | 7 | 1186 |