Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000371.3 | NP_000362.1 | 3 | 390 | ||
GenBank mRNA | AF162690.1 | - | 3 | 259 | ||
AK312051.1 | - | 3 | 282 | |||
BC005310.1 | - | 3 | 280 | 4071761 | ||
BC020791.1 | - | 3 | 280 | 4734010 | ||
BT007189.1 | - | 3 | 254 | |||
CR456908.1 | - | 3 | 254 | |||
D00096.1 | - | 3 | 280 | |||
DB498675.1 | - | 3 | 390 | |||
DQ839490.1 | - | 3 | 254 | |||
EU794670.1 | - | 3 | 254 | |||
GU727633.1 | - | 3 | 280 | |||
K02091.1 | - | 3 | 280 | |||
M10605.1 | - | 3 | 279 | |||
M11714.1 | - | 2 | 166 | |||
U19780.1 | - | 3 | 254 | |||
X59498.1 | - | 3 | 275 |