Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030203.2 | NP_084479.1 | 1 | 530 | ||
GenBank mRNA | AK020999.1 | - | 1 | 441 | ||
AK030740.1 | - | 1 | 475 | |||
AK034070.1 | - | 1 | 497 | |||
AK034414.1 | - | 1 | 471 | |||
AK158197.1 | - | 1 | 498 | |||
BC017540.1 | - | 1 | 479 | 4511620 | ||
BC034656.1 | - | 1 | 478 | 5361000 |