Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030205.4 | NP_084481.3 | 2, 3 | 274 | ||
GenBank mRNA | AK018739.1 | - | 2, 3 | 242 | ||
AK037894.1 | - | 2, 3 | 268 | |||
AK039585.1 | - | 2, 3 | 331 | |||
AK040408.1 | - | 280 | ||||
AK088155.1 | - | 2, 3 | 242 | |||
AK143624.1 | - | 2, 3 | 233 | |||
AK150051.1 | - | 297 | ||||
AK178516.1 | - | 1, 2 | 68 | |||
AK180866.1 | - | 15 | ||||
AK214435.1 | - | 20 | ||||
BC061006.1 | - | 2, 3 | 274 | 6536106 |