Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030215.3 | NP_084491.3 | 2 | 1073 | ||
GenBank mRNA | AB056151.1 | - | 2 | 922 | ||
AF208046.1 | - | 2 | 318 | |||
AK041886.1 | - | 2 | 1029 | |||
AK050368.1 | - | 2 | 1075 | |||
AK082078.1 | - | 2 | 988 | |||
AK167570.1 | - | 2 | 1074 | |||
AK170542.1 | - | 2 | 1066 | |||
AK170846.1 | - | 2 | 1054 | |||
AK197211.1 | - | 1 | 65 | |||
AK210446.1 | - | 1 | 65 | |||
BC010482.1 | - | 2 | 1065 | 3492125 | ||
BC058744.1 | - | 2 | 463 |