Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030229.4 | NP_084505.2 | 1 | 409 | ||
GenBank mRNA | AK019868.2 | - | 1 | 410 | ||
AK032967.1 | - | 361 | ||||
AK152517.1 | - | 1 | 382 | |||
BC010793.1 | - | 1 | 412 | 4221321 |