Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030239.2 | NP_084515.2 | 2 | 298 | ||
GenBank mRNA | AF324242.1 | - | 2 | 250 | ||
AK030831.1 | - | 2 | 300 | |||
AK037614.1 | - | 2 | 214 | |||
AK051880.1 | - | 2 | 264 | |||
AK156626.1 | - | 2 | 266 | |||
BC029095.1 | - | 1 | 143 | |||
BC138772.1 | - | 2 | 279 | 8861794 | ||
BC138773.1 | - | 2 | 278 | 8861795 |