Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030252.2 | NP_084528.1 | 2 | 347 | ||
GenBank mRNA | AK075968.1 | - | 2 | 322 | ||
AK131915.1 | - | 2 | 312 | |||
AK141221.1 | - | 2 | 281 | |||
BC147067.1 | - | 2 | 309 | 9056070 | ||
BC147068.1 | - | 2 | 309 | 9056074 | ||
BY237825.1 | - | 2 | 348 |