Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160415.1 | NP_001153887.1 | 2, 3 | 318 | ||
NM_030255.3 | NP_084531.2 | 2, 3 | 318 | |||
GenBank mRNA | AB461446.1 | - | 2, 3 | 209 | ||
AB461447.1 | - | 2, 3 | 209 | |||
AB461448.1 | - | 2, 3 | 209 | |||
AB461449.1 | - | 2, 3 | 209 | |||
AK044394.1 | - | 2, 3 | 278 | |||
AK049256.1 | - | 2, 3 | 279 | |||
AK049998.1 | - | 272 | ||||
AK153719.1 | - | 2, 3 | 268 | |||
AK154035.1 | - | 323 | ||||
AK167986.1 | - | 2, 3 | 324 | |||
BC003314.1 | - | 2, 3 | 300 | 3155422 | ||
BY228556.1 | - | 2, 3 | 320 | |||
EU707568.1 | - | 2, 3 | 209 | |||
EU707570.1 | - | 2, 3 | 209 | |||
GQ871501.1 | - | 2, 3 | 206 | |||
GQ871502.1 | - | 1, 2 | 149 | |||
GQ871503.1 | - | 2, 3 | 206 |