Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pofut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030262.3 | NP_084538.2 | 1 | 98 | ||
GenBank mRNA | AF455270.1 | - | 1 | 88 | ||
AK009301.1 | - | 1 | 97 | |||
AK132373.1 | - | 1 | 95 | |||
AK147414.1 | - | 1 | 80 | |||
AK148244.1 | - | 1 | 97 | |||
AK194758.1 | - | 1 | 85 | |||
AK198059.1 | - | 1 | 23 | |||
BC018194.1 | - | 1 | 96 | |||
BC147077.1 | - | 1 | 165 | 9056064 | ||
BC147078.1 | - | 1 | 165 | 9056063 | ||
CO814561.1 | - | 98 |