Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159369.1 | NP_001152841.1 | 1, 2 | 282 | ||
NM_029977.2 | NP_084253.1 | 1, 2 | 282 | |||
GenBank mRNA | AK020790.1 | - | 1, 2 | 253 | ||
AY074936.1 | - | 1, 2 | 282 | |||
AY147862.1 | - | 1, 2 | 282 | |||
AY147863.1 | - | 1, 2 | 282 | |||
AY147864.1 | - | 1, 2 | 282 | |||
BC138360.1 | - | 1, 2 | 223 | 8861380 | ||
BC157900.1 | - | 1, 2 | 223 | 9088092 |