Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029999.4 | NP_084275.3 | 3 | 520 | ||
GenBank mRNA | AF317517.1 | - | 3 | 503 | ||
AK020110.1 | - | 3 | 520 | |||
BC026827.1 | - | 3 | 502 | 4218560 | ||
BC052470.1 | - | 3 | 500 | 6813866 |