Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030004.3 | NP_084280.2 | 3 | 196 | ||
GenBank mRNA | AB359906.1 | - | 3 | 161 | ||
AF351609.1 | - | 3 | 241 | |||
AK003913.1 | - | 3 | 235 | |||
AK020717.1 | - | 3 | 210 | |||
AK043569.1 | - | 3 | 243 | |||
AK080625.1 | - | 3 | 217 | |||
AK135834.1 | - | 3 | 210 | |||
AK207089.1 | - | 3 | 141 | |||
AK218806.1 | - | 3 | 141 | |||
BC004074.1 | - | 3 | 196 | 3591359 | ||
BC027064.1 | - | 3 | 213 | 5355916 |