Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291125.1 | NP_001278054.1 | 3 | 358 | ||
NM_001291126.1 | NP_001278055.1 | 3 | 189 | |||
NM_001291127.1 | NP_001278056.1 | 2 | 205 | |||
NM_030064.4 | NP_084340.2 | 3 | 358 | |||
GenBank mRNA | AK019135.1 | - | 3 | 346 | ||
AK029376.1 | - | 3 | 139 | |||
AK031159.1 | - | 3 | 107 | |||
AK170022.1 | - | 2 | 206 | |||
AK170729.1 | - | 3 | 275 | |||
AY099326.1 | - | 3 | 104 | |||
AY099327.1 | - | 3 | 104 | |||
BC033533.1 | - | 3 | 248 | 2655211 | ||
BM947867.1 | - | 2 | 128 | |||
BY262829.1 | - | 3 | 166 | |||
BY294224.1 | - | 3 | 353 | |||
CA750740.1 | - | 3 | 338 | |||
DV662694.1 | - | 3 | 317 |