Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282126.1 | NP_001269055.1 | 2 | 715 | ||
NM_001282127.1 | NP_001269056.1 | 2 | 483 | |||
NM_001282128.1 | NP_001269057.1 | 2 | 715 | |||
NM_030178.2 | NP_084454.1 | 2 | 483 | |||
GenBank mRNA | AK043372.1 | - | 2 | 716 | ||
AK156375.1 | - | 2 | 663 | |||
AK206283.1 | - | 1 | 192 | |||
BC046521.1 | - | 2 | 423 | 5363697 | ||
BC138361.1 | - | 2 | 307 | 8861381 | ||
BC145256.1 | - | 2 | 539 | 9053809 | ||
DQ288860.1 | - | 1 | 272 |