Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspyl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030203.2 | NP_084479.1 | 1 | 623 | ||
GenBank mRNA | AK030740.1 | - | 1 | 568 | ||
AK034070.1 | - | 1 | 590 | |||
AK034414.1 | - | 1 | 564 | |||
AK158197.1 | - | 1 | 591 | |||
BC017540.1 | - | 1 | 572 | 4511620 | ||
BC030922.1 | - | 1 | 98 | 4527889 | ||
BC034656.1 | - | 1 | 571 | 5361000 |