Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030245.3 | NP_084521.1 | 2, 3 | 403 | ||
GenBank mRNA | AK013599.1 | - | 2, 3 | 170 | ||
AK050649.1 | - | 2, 3 | 247 | |||
AK151132.1 | - | 2, 3 | 265 | |||
BC002307.1 | - | 2, 3 | 205 | 3592046 | ||
BC027337.1 | - | 2, 3 | 270 | 3995355 |