Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB461446.1 | - | 3 | 341 | ||
AB461447.1 | - | 3 | 341 | |||
AB461448.1 | - | 3 | 341 | |||
AK044394.1 | - | 3 | 410 | |||
AK154035.1 | - | 455 | ||||
BC003314.1 | - | 3 | 432 | 3155422 | ||
EU707569.1 | - | 2 | 173 | |||
EU707571.1 | - | 2 | 173 | |||
GQ871501.1 | - | 3 | 338 | |||
GQ871502.1 | - | 2 | 281 | |||
GQ871503.1 | - | 3 | 338 |