Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030259.1 | NP_084535.1 | 1 | 397 | ||
GenBank mRNA | AK133760.1 | - | 1 | 529 | ||
AK150123.1 | - | 1 | 501 | |||
AK150252.1 | - | 1 | 500 | |||
AK171419.1 | - | 1 | 475 | |||
BC003324.1 | - | 1 | 397 | 3156139 |