Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029977.2 | NP_084253.1 | 7 | 1124 | ||
GenBank mRNA | AY074936.1 | - | 7 | 1124 | ||
AY147863.1 | - | 8 | 1220 | |||
BC138360.1 | - | 7 | 1065 | 8861380 | ||
BC157900.1 | - | 7 | 1065 | 9088092 |