Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291125.1 | NP_001278054.1 | 5 | 1711 | ||
NM_001291126.1 | NP_001278055.1 | 5 | 1743 | |||
NM_001291127.1 | NP_001278056.1 | 4 | 1759 | |||
NM_030064.4 | NP_084340.2 | 5 | 1912 | |||
GenBank mRNA | AK019135.1 | - | 5 | 1900 | ||
AK029376.1 | - | 5 | 1693 | |||
AK031159.1 | - | 5 | 1661 | |||
AK170022.1 | - | 4 | 1758 | |||
BC033533.1 | - | 5 | 1601 | 2655211 | ||
BY632579.1 | - | 1 | 298 |