Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030199.3 | NP_084475.1 | 2 | 1593 | ||
GenBank mRNA | AK019163.1 | - | 2 | 1490 | ||
AK028561.1 | - | 2 | 1414 | |||
AK030476.1 | - | 2 | 1498 | |||
AK046806.1 | - | 2 | 1594 | |||
AK169386.1 | - | 2 | 1591 | |||
AK172985.1 | - | 2 | 1568 | |||
AY610936.1 | - | 2 | 1594 | |||
BC021834.1 | - | 2 | 1496 | 4022529 | ||
BC086767.1 | - | 2 | 1484 | 6832277 |