Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pagr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030240.1 | NP_084516.1 | 3 | 1045 | ||
GenBank mRNA | AK013840.1 | - | 3 | 1019 | ||
AK082775.1 | - | 3 | 754 | |||
BC003932.1 | - | 3 | 1045 | 3485982 | ||
BC054744.1 | - | 4182 |