Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030250.2 | NP_084526.1 | 5 | 1126 | ||
GenBank mRNA | AK162260.1 | - | 3 | 288 | ||
AK167741.1 | - | 5 | 1115 | |||
AK167939.1 | - | 5 | 1119 | |||
BC003223.1 | - | 5 | 1085 | 3482163 | ||
BC018372.1 | - | 5 | 1101 | 3499868 |