Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030251.3 | NP_084527.2 | 7, 8 | 749 | ||
GenBank mRNA | AB053477.1 | - | 7, 8 | 680 | ||
AK049100.1 | - | 7, 8 | 665 | |||
AK088437.1 | - | 7, 8 | 714 | |||
AK089474.1 | - | 5, 6 | 1113 | |||
AK155432.1 | - | 2, 3 | 2171 | |||
AK155505.1 | - | 2, 3 | 2171 | |||
AK155644.1 | - | 2, 3 | 2171 | |||
BC003234.1 | - | 7, 8 | 693 | 3483893 |