Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163348.1 | NP_001156820.1 | 2 | 1227 | ||
NM_001163349.1 | NP_001156821.1 | 2 | 1227 | |||
NM_001163350.1 | NP_001156822.1 | 2 | 1227 | |||
NM_001163351.1 | NP_001156823.1 | 2 | 1227 | |||
NM_030699.2 | NP_109624.1 | 2 | 1227 | |||
NM_133488.1 | NP_597995.1 | 2 | 1227 | |||
GenBank mRNA | AB038662.1 | - | 1 | 132 | ||
AB038663.1 | - | 1 | 132 | |||
AB038664.1 | - | 1 | 132 | |||
AB038665.1 | - | 1 | 132 | |||
AB038666.1 | - | 1 | 132 | |||
AB038667.1 | - | 2 | 1004 | |||
AF475069.1 | - | 1 | 132 | |||
AF475070.1 | - | 1 | 132 | |||
AF475071.1 | - | 1 | 132 | |||
AF475072.1 | - | 1 | 132 | |||
AF475073.1 | - | 1 | 132 | |||
AF475074.1 | - | 1 | 132 | |||
AF475075.1 | - | 1 | 132 | |||
AF475076.1 | - | 1 | 132 | |||
AF475077.1 | - | 1 | 132 | |||
AF475078.1 | - | 1 | 132 | |||
AK034465.1 | - | 2 | 761 | |||
AK048796.1 | - | 1 | 93 | |||
AK134467.1 | - | 2 | 1230 | |||
AK173075.1 | - | 2 | 1012 | |||
BC079881.1 | - | 2 | 1194 | 6837530 |