Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030714.2 | NP_109639.1 | 4 | 586 | ||
GenBank mRNA | AB015425.1 | - | 3 | 568 | ||
AK148444.1 | - | 4 | 605 | |||
BC099687.1 | - | 4 | 586 | 6311735 | ||
BC138304.1 | - | 3 | 465 | 8861323 | ||
BC157899.1 | - | 3 | 467 | 9088091 |