Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030715.3 | NP_109640.1 | 2 | 332 | ||
GenBank mRNA | AB027128.1 | - | 2 | 358 | ||
AK036296.1 | - | 2 | 333 | |||
BC049159.1 | - | 2 | 303 | |||
BC128366.1 | - | 2 | 329 | 40111620 |