Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000416.2 | NP_000407.1 | 2, 3 | 297 | ||
XM_006715470.2 | XP_006715533.1 | 3, 4 | 658 | |||
XM_006715471.2 | XP_006715534.1 | 2, 3 | 547 | |||
XM_011535793.2 | XP_011534095.1 | 3, 4 | 1329 | |||
XM_011535794.1 | XP_011534096.1 | 3, 4 | 274 | |||
XM_017010827.1 | XP_016866316.1 | 3, 4 | 368 | |||
GenBank mRNA | AK222803.1 | - | 2, 3 | 252 | ||
AK294252.1 | - | 3, 4 | 340 | |||
AK297052.1 | - | 2, 3 | 173 | |||
AK312674.1 | - | 2, 3 | 251 | |||
BC005333.1 | - | 2, 3 | 276 | 3950528 | ||
BT006814.1 | - | 2, 3 | 194 | |||
DC349115.1 | - | 2, 3 | 297 | |||
EF535103.1 | - | 2, 3 | 237 | |||
J03143.1 | - | 2, 3 | 242 | |||
KU178144.1 | - | 2, 3 | 194 | |||
KU178145.1 | - | 2, 3 | 194 |