Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030750.3 | NP_109675.1 | 1 | 726 | ||
GenBank mRNA | AF247177.1 | - | 1 | 676 | ||
AF415215.1 | - | 1 | 560 | |||
AK043586.1 | - | 1 | 694 | |||
AK149860.1 | - | 1 | 727 | |||
AK154116.1 | - | 2 | 368 | |||
BC026814.1 | - | 1 | 662 | |||
BC037592.1 | - | 1 | 679 | 5004335 |