Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC004004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030561.3 | NP_085038.3 | 3, 4 | 522 | ||
GenBank mRNA | AK030596.1 | - | 1, 2 | 92 | ||
AK217020.1 | - | 2, 3 | 312 | |||
BC004004.1 | - | 3, 4 | 359 | 3492768 | ||
BC058575.1 | - | 3, 4 | 331 | 6488611 | ||
CD353313.1 | - | 3, 4 | 474 |