Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030566.2 | NP_085043.2 | 3, 4 | 766 | ||
GenBank mRNA | AK089938.1 | - | 3, 4 | 470 | ||
AK144836.1 | - | 3, 4 | 452 | |||
AK157832.1 | - | 3, 4 | 427 | |||
BC004088.1 | - | 2, 3 | 512 | 3593722 | ||
BC015287.1 | - | 3, 4 | 774 | 4240442 |