Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286653.1 | NP_001273582.1 | 2, 3 | 348 | ||
NM_001286654.1 | NP_001273583.1 | 2, 3 | 225 | |||
NM_030598.2 | NP_085101.1 | 1, 2 | 355 | |||
NM_207649.1 | NP_997532.1 | 2, 3 | 481 | |||
GenBank mRNA | AB061524.1 | - | 1, 2 | 293 | ||
AB061525.1 | - | 2, 3 | 348 | |||
AF237791.1 | - | 1, 2 | 95 | |||
AF237887.1 | - | 1, 2 | 141 | |||
AK049006.1 | - | 1, 2 | 294 | |||
AK136805.1 | - | 2, 3 | 289 | |||
AK138768.1 | - | 2, 3 | 393 | |||
AK160865.1 | - | 1, 2 | 243 | |||
BB635249.1 | - | 2, 3 | 393 | |||
BC047153.1 | - | 1, 2 | 153 | 6491318 | ||
BC049096.1 | - | 1, 2 | 355 | 6416901 | ||
BC062141.1 | - | 2, 3 | 225 | 30290791 | ||
BF534085.1 | - | 328 | ||||
BY740155.1 | - | 2, 3 | 488 |