Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030688.1 | NP_109613.1 | 3 | 369 | ||
GenBank mRNA | AF284437.1 | - | 3 | 369 | ||
AJ277831.1 | - | 1127 | ||||
AK039155.1 | - | 894 | ||||
AK045976.1 | - | 1158 | ||||
BC119274.1 | - | 3 | 312 | 8734023 | ||
BC120555.1 | - | 3 | 312 | 40129478 |