Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000422.2 | NP_000413.1 | 4 | 825 | ||
GenBank mRNA | AK095342.1 | - | 4 | 754 | ||
BC000159.2 | - | 4 | 754 | 2986100 | ||
BC011901.2 | - | 4 | 752 | 4111703 | ||
BC056421.1 | - | 4 | 754 | 5208304 | ||
BC072018.1 | - | 5 | 692 | 3531173 | ||
BC072019.1 | - | 4 | 750 | 5442690 | ||
BX647923.1 | - | 3 | 1091 | |||
X05803.1 | - | 3 | 206 | |||
X62571.1 | - | 4 | 752 |